Anti-SACS Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A68554-100UG A68554-50UG
77838-941EA 330 USD
77838-941 77838-942
Anti-SACS Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: SACS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR