Anti-R3HDML Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A68242-100UG A68242-50UG
77838-355EA 330 USD
77838-355 77838-356
Anti-R3HDML Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: R3HDML
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR