Anti-GLO1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A66866-100UL A66866-50UL
77835-771EA 330 USD
77835-771 77835-772
Anti-GLO1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: GLO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR