Anti-GT-1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A66673-100UG A66673-50UG
77835-408EA 330 USD
77835-408 77835-409
Anti-GT-1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: GT-1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR