Anti-KRT12 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A66355-100UL A66355-50UL
77834-811EA 330 USD
77834-811 77834-812
Anti-KRT12 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: KRT12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR