Anti-P2RX7 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A66307-100UL A66307-50UL
77834-718EA 330 USD
77834-718 77834-719
Anti-P2RX7 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: P2RX7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR