Anti-FAM118B Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A66206-100UG A66206-50UG
77834-530EA 330 USD
77834-530 77834-531
Anti-FAM118B Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: FAM118B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR