Anti-FLAD1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A65647-100UG A65647-50UG
77833-471EA 330 USD
77833-471 77833-472
Anti-FLAD1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200

Type:
Antigen: FLAD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR