Anti-FAM111A Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A65497-100UG A65497-50UG
77833-184EA 330 USD
77833-184 77833-185
Anti-FAM111A Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: FAM111A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR