Anti-TMPRSS11B Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60006-100UL A60006-50UL
77832-940EA 330 USD
77832-940 77832-941
Anti-TMPRSS11B Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:1000 to 1:2000, IHC:1:10 to 1:50

Type:
Antigen: TMPRSS11B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR