Anti-UBE2B Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A59974-100UL A59974-50UL
77832-890EA 330 USD
77832-890 77832-891
Anti-UBE2B Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100

Type:
Antigen: UBE2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR