Anti-LYZL2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A59758-100UG A59758-50UG
77832-497EA 330 USD
77832-497 77832-498
Anti-LYZL2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: LYZL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR