Anti-KTN1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A59681-100UG A59681-50UG
77832-351EA 330 USD
77832-351 77832-352
Anti-KTN1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: KTN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR