Anti-ARMCX6 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A59276-100UG A59276-50UG
77831-587EA 330 USD
77831-587 77831-588
Anti-ARMCX6 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: ARMCX6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR