Anti-C8orf74 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A59166-100UG A59166-50UG
77831-382EA 330 USD
77831-382 77831-383
Anti-C8orf74 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: C8orf74
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR