Anti-FGD6 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A59101-100UG A59101-50UG
77831-260EA 330 USD
77831-260 77831-261
Anti-FGD6 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: FGD6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR