Anti-SLC24A5 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A58827-100UG A58827-50UG
77830-750EA 330 USD
77830-750 77830-751
Anti-SLC24A5 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SLC24A5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR