Anti-REG3G Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A58620-100UG A58620-50UG
77830-354EA 330 USD
77830-354 77830-355
Anti-REG3G Rabbit Recombinant Antibody
Antibodies
Type:
Antigen: REG3G
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR