Anti-TCTN3 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A58444-100UG A58444-50UG
77830-020EA 330 USD
77830-020 77830-021
Anti-TCTN3 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TCTN3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR