Anti-ATG1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A58060-100UG A58060-50UG
77829-292EA 330 USD
77829-292 77829-293
Anti-ATG1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: ATG1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Candida glabrata
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR