Anti-CCDC181 Rabbit Recombinant Antibody (Biotinylated)

Supplier: AFG Bioscience
A57698-50UG A57698-100UG
77828-612EA 225 USD
77828-612 77828-611
Anti-CCDC181 Rabbit Recombinant Antibody (Biotinylated)
Antibodies
Type:
Antigen: CCDC181
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR