Anti-Conglutin Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A57533-100UG A57533-50UG
77828-305EA 330 USD
77828-305 77828-306
Anti-Conglutin Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Conglutin
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Peanut
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR