Anti-HYI Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A57475-100UG A57475-50UG
77828-193EA 330 USD
77828-193 77828-194
Anti-HYI Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: HYI
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR