Anti-KIF24 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A57369-100UG A57369-50UG
77827-993EA 330 USD
77827-993 77827-994
Anti-KIF24 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: KIF24
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR