Anti-ompA Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A57160-100UG A57160-50UG
77827-603EA 330 USD
77827-603 77827-604
Anti-ompA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: ompA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rickettsia conorii
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR