Anti-ASB10 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A56874-100UL A56874-50UL
77827-098EA 330 USD
77827-098 77827-099
Anti-ASB10 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000

Type:
Antigen: ASB10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR