Anti-FREM1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A56764-100UG A56764-50UG
77826-887EA 330 USD
77826-887 77826-888
Anti-FREM1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:500 to 1:1000

Type:
Antigen: FREM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR