Anti-TKFC Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A56339-100UG A56339-50UG
77826-086EA 330 USD
77826-086 77826-087
Anti-TKFC Rabbit Recombinant Antibody
Antibodies
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200

Type:
Antigen: TKFC
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR