Anti-AOC3 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A56042-100UG A56042-50UG
77825-533EA 330 USD
77825-533 77825-534
Anti-AOC3 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: AOC3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR