Anti-EIF4EBP1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A55887-100UG A55887-50UG
77825-234EA 330 USD
77825-234 77825-235
Anti-EIF4EBP1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: EIF4EBP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR