Anti-LAMA3 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A55815-100UG A55815-50UG
77825-099EA 330 USD
77825-099 77825-100
Anti-LAMA3 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: LAMA3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR