Anti-APOBEC2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A55714-100UL A55714-50UL
77824-909EA 330 USD
77824-909 77824-910
Anti-APOBEC2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000

Type:
Antigen: APOBEC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR