Anti-LAMA4 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A55543-100UL A55543-50UL
77824-577EA 330 USD
77824-577 77824-578
Anti-LAMA4 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: LAMA4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR