Anti-STK4 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A55386-100UL A55386-50UL
77824-278EA 330 USD
77824-278 77824-279
Anti-STK4 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: STK4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR