Anti-PCK2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A55128-100UG A55128-50UG
77823-792EA 330 USD
77823-792 77823-793
Anti-PCK2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: PCK2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR