Anti-TRIP4 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A55065-100UG A55065-50UG
77823-672EA 330 USD
77823-672 77823-673
Anti-TRIP4 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TRIP4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR