Anti-PCM1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A55040-100UL A55040-50UL
77823-626EA 330 USD
77823-626 77823-627
Anti-PCM1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: PCM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR