Anti-TP73 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A54680-100UL A54680-50UL
77822-959EA 330 USD
77822-959 77822-960
Anti-TP73 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000

Type:
Antigen: TP73
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR