Anti-OVGP1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A54352-100UG A54352-50UG
77822-333EA 330 USD
77822-333 77822-334
Anti-OVGP1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: OVGP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR