Anti-RBMS2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A54256-100UG A54256-50UG
77822-152EA 330 USD
77822-152 77822-153
Anti-RBMS2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: RBMS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR