Anti-UNC79 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A53957-100UG A53957-50UG
77821-579EA 330 USD
77821-579 77821-580
Anti-UNC79 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: UNC79
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR