Anti-EHHADH Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A53921-100UG A53921-50UG
77821-508EA 330 USD
77821-508 77821-509
Anti-EHHADH Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: EHHADH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR