Anti-AQP2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A53856-100UL A53856-50UL
77821-396EA 330 USD
77821-396 77821-397
Anti-AQP2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:2000 to 1:10000, IHC: 1:100 to 1:300

Type:
Antigen: AQP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR