Anti-TRAF3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A53804-100UL A53804-50UL
77821-309EA 330 USD
77821-309 77821-310
Anti-TRAF3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200

Type:
Antigen: TRAF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR