Anti-ALB Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A53766-100UL A53766-50UL
77821-248EA 330 USD
77821-248 77821-249
Anti-ALB Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:10000, WB: 1:1000 to 1:5000

Type:
Antigen: ALB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR