Anti-IgG(H+L) Goat Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A53211-100UL A53211-500UL
77820-345EA 90 USD
77820-345 77820-346
Anti-IgG(H+L) Goat Polyclonal Antibody (HRP)
Antibodies
Recommended dilution: ELISA: 1:500 to 1:10000, WB: 1:3000 to 1:5000, IHC: 1:50 to 1:500, ICC: 1:50 to 1:500

Type: Secondary
Antigen: IgG(H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR