Anti-CLK2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A53154-100UL
77820-251EA 330 USD
77820-251
Anti-CLK2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:3000

Type:
Antigen: CLK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR