Anti-ACPP Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A52968-100UL A52968-50UL
77819-929EA 330 USD
77819-929 77819-930
Anti-ACPP Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:200 to 1:1000

Type:
Antigen: ACPP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR