Anti-SHISA9 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A52348-100UL A52348-50UL
77818-800EA 330 USD
77818-800 77818-801
Anti-SHISA9 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: SHISA9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR