Anti-HNF4A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A52190-100UL
77818-538EA 399 USD
77818-538
Anti-HNF4A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:1000

Type:
Antigen: HNF4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR